2-Bromo-6-pyridinemethanol, 96%
2-Bromo-6-pyridinemethanol, 96%
2-Bromo-6-pyridinemethanol, 96%
Thermo Scientific Chemicals

2-Bromo-6-pyridinemethanol, 96%

CAS: 33674-96-3 | C6H6BrNO | 188.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H56123.03
also known as H56123-03
Price (USD)
59.65
Online Exclusive
66.50
Save 6.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
66.50
Save 6.85 (10%)
Each
Chemical Identifiers
CAS33674-96-3
IUPAC Name(6-bromopyridin-2-yl)methanol
Molecular FormulaC6H6BrNO
InChI KeyXDDGKNRSCDEWBR-UHFFFAOYSA-N
SMILESOCC1=CC=CC(Br)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)96%
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and bases. Store at room temperature.
RUO – Research Use Only
Marta Martínez Belmonte.; Eduardo C. Escudero-Adán.; Jordi Benet-Buchholz.; Arjan W. Kleij. Formation of Unusual Trinuclear Assemblies: Scope and Mechanism of Zn(salphen)-Templated Activation of Pyridine-Alcohol Substrates. Eur. J. Inorg. Chem. 2009, 2009 (35),5307-5318.