5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98+%, Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98+%, Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H56265.03
also known as H56265-03
1 g
Catalog number H56265.03
also known as H56265-03
Price (USD)
307.65
Online Exclusive
342.00
Save 34.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
307.65
Online Exclusive
342.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS102185-33-1
IUPAC Namedisodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
Molecular FormulaC8H4BrClNNa2O4P
InChI KeyOAZUOCJOEUNDEK-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
It is an histochemical substrate for alkaline phosphatase. For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an histochemical substrate for alkaline phosphatase. For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

Solubility
Soluble in water and toluene.

Notes
Moisture and light sensitive. Store in dry place. Keep away from oxidizing agents, heat and light.
RUO – Research Use Only
Chuanmin Ruan.; Liju Yang.; Yanbin Li. Immunobiosensor Chips for Detection of Escherichia coli O157:H7 Using Electrochemical Impedance Spectroscopy. Anal. Chem. 2002, 74 (18), 4814-4820 .