(S)-(+)-3-Hydroxytetrahydrofuran, 98%
(S)-(+)-3-Hydroxytetrahydrofuran, 98%
(S)-(+)-3-Hydroxytetrahydrofuran, 98%
Thermo Scientific Chemicals

(S)-(+)-3-Hydroxytetrahydrofuran, 98%

CAS: 86087-23-2 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H56476.06
also known as H56476-06
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.60
Save 8.95 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.60
Save 8.95 (15%)
Each
Chemical Identifiers
CAS86087-23-2
IUPAC Nameoxolan-3-ol
Molecular FormulaC4H8O2
InChI KeyXDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILESOC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only