Sodium 4-hydroxybenzoate, 99+%
Sodium 4-hydroxybenzoate, 99+%
Sodium 4-hydroxybenzoate, 99+%
Sodium 4-hydroxybenzoate, 99+%
Thermo Scientific Chemicals

Sodium 4-hydroxybenzoate, 99+%

CAS: 114-63-6 | C7H5NaO3 | 160.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Catalog number H56590.22
also known as H56590-22
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS114-63-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>99.0%
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
Line Ste-Marie.; Daniel Boismenu.; Luc Vachon.; Jane Montgomery. Evaluation of Sodium 4-Hydroxybenzoate as an Hydroxyl Radical Trap Using Gas Chromatography-Mass Spectrometry and High-Performance Liquid Chromatography with Electrochemical Detection. Anal. Biochem. 1996, 241 (1),67-74 .