4-Chloro-7-azaindole, 98%
4-Chloro-7-azaindole, 98%
4-Chloro-7-azaindole, 98%
4-Chloro-7-azaindole, 98%
Thermo Scientific Chemicals

4-Chloro-7-azaindole, 98%

CAS: 55052-28-3 | C7H5ClN2 | 152.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H56734.03
also known as H56734-03
Price (USD)
86.65
Special offer
Online exclusive
Ends: 31-Dec-2025
102.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 31-Dec-2025
102.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS55052-28-3
IUPAC Name4-chloro-1H-pyrrolo[2,3-b]pyridine
Molecular FormulaC7H5ClN2
InChI KeyHNTZVGMWXCFCTA-UHFFFAOYSA-N
SMILESClC1=C2C=CNC2=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Assay (HPLC)>97.5%
FormSolid
A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. John J. Caldwell, et al. Synthesis of 4-(cyclic dialkylamino)-7-azaindoles by microwave heating of 4-halo-7-azaindoles and cyclic secondary amines.Tetrahedron Lett.,2007,48(9), 1527-1529.
  2. Jérome Guillard, et al. Synthesis and biological evaluation of 7-azaindole derivatives, synthetic cytokinin analogues.Bioorg. Med. Chem. Lett.,2007,17(7), 1934-1937.