Bromomethyl acetate, 95%
Bromomethyl acetate, 95%
Bromomethyl acetate, 95%
Thermo Scientific Chemicals

Bromomethyl acetate, 95%

CAS: 590-97-6 | C3H5BrO2 | 152.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number H56755.06
also known as H56755-06
Price (USD)
283.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
283.00
Each
Chemical Identifiers
CAS590-97-6
IUPAC Namebromomethyl acetate
Molecular FormulaC3H5BrO2
InChI KeyNHYXMAKLBXBVEO-UHFFFAOYSA-N
SMILESCC(=O)OCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)95%
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

Solubility
Miscible with chloroform.

Notes
Store in cool place. Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Pansuriya, P. B.; Parekh, H. M.; Maguire, G. E. M.; Friedrich, H. B. Tetramethoxy resorcin[4]arene-tetraester derivatives. J. Therm. Anal. Calorim. 2015, 120 (1), 653-665.
  2. Neuenschwander, M. Low-Temperature Olefin Syntheses in View of Parent Fulvenes and Fulvalenes. Helv. Chim. Acta 2015, 98 (6), 731-762.