2,4,6-Trichloropyridine, 97%
2,4,6-Trichloropyridine, 97%
2,4,6-Trichloropyridine, 97%
Thermo Scientific Chemicals

2,4,6-Trichloropyridine, 97%

CAS: 16063-69-7 | C5H2Cl3N | 182.428 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H56787.14
also known as H56787-14
Price (USD)
465.65
Online Exclusive
517.00
Save 51.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
465.65
Online Exclusive
517.00
Save 51.35 (10%)
Each
Chemical Identifiers
CAS16063-69-7
IUPAC Name2,4,6-trichloropyridine
Molecular FormulaC5H2Cl3N
InChI KeyFJNNGKMAGDPVIU-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0%
FormSolid
It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Sparingly soluble in water. (0.113 mm Hg at 25°C)

Notes
Store at 2-8°C. Avoid using oxidizing agents.
RUO – Research Use Only

General References:

  1. Manfred Schlosser.; Thierry Rausis.; Carla Bobbio. Rerouting Nucleophilic Substitution from the 4-Position to the 2- or 6-Position of 2,4-Dihalopyridines and 2,4,6-Trihalopyridines: The Solution to a Long-Standing Problem.Org. Lett. 2005, 7 (1),127-129.
  2. Jesse V. Mello.; Nathaniel S. Finney. Convenient Synthesis and Transformation of 2,6-Dichloro-4-iodopyridine. Org. Lett. 2001, 3 (26),4263-4265.