(+/-)-1-Boc-3-bromopyrrolidine, 95%
(+/-)-1-Boc-3-bromopyrrolidine, 95%
(+/-)-1-Boc-3-bromopyrrolidine, 95%
Thermo Scientific Chemicals

(+/-)-1-Boc-3-bromopyrrolidine, 95%

CAS: 939793-16-5 | C9H16BrNO2 | 250.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H57071.06
also known as H57071-06
Price (USD)
453.65
Special offer
Online exclusive
Ends: 30-Jun-2026
534.00
Save 80.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
453.65
Special offer
Online exclusive
Ends: 30-Jun-2026
534.00
Save 80.35 (15%)
Each
Specifications
CAS939793-16-5
Chemical Name or Material(+/-)-1-Boc-3-bromopyrrolidine
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Formula Weight250.14
Health Hazard 1H302-H319
View more
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only