2-Chloro-N,N-dimethylnicotinamide, 97%
2-Chloro-N,N-dimethylnicotinamide, 97%
2-Chloro-N,N-dimethylnicotinamide, 97%
2-Chloro-N,N-dimethylnicotinamide, 97%
Thermo Scientific Chemicals

2-Chloro-N,N-dimethylnicotinamide, 97%

CAS: 52943-21-2 | C8H9ClN2O | 184.623 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H57162.03
also known as H57162-03
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
126.00
Save 18.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
126.00
Save 18.35 (15%)
Each
Add to cart
Specifications
CAS52943-21-2
Chemical Name or Material2-Chloro-N,N-dimethylnicotinamide
Formula Weight184.63
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only