DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals
DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals
DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57225.03
also known as H57225-03
1 g
Catalog number H57225.03
also known as H57225-03
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Specifications
Beilstein1420904
Boiling Point165°C to 171°C
CAS5082-74-6
Chemical Name or MaterialDL-beta-Prolinol
Density0.979
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
RUO – Research Use Only
David Alker.; Kevin J. Doyle.; Laurence M. Harwood.; Andrew McGregor. The direct synthesis of the cyclic sulphamidate of (S)-prolinol: SimultaneousN-protection and activation towards nucleophilic displacement of oxygen. Tetrahedron: Asymmetry. 1990, 1 (12),877-880.