3-Bromomethyl-3-oxetanemethanol, 95%
3-Bromomethyl-3-oxetanemethanol, 95%
3-Bromomethyl-3-oxetanemethanol, 95%
Thermo Scientific Chemicals

3-Bromomethyl-3-oxetanemethanol, 95%

CAS: 22633-44-9 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57413.MD
also known as H57413-MD
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
56.30
Save 8.65
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
56.30
Save 8.65
Each
Specifications
CAS22633-44-9
Chemical Name or Material3-Bromomethyl-3-oxetanemethanol
Density1.598
Flash Point100°C (212°F)
Formula Weight181.03
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only