3-(Methylamino)oxetane, 95%
3-(Methylamino)oxetane, 95%
3-(Methylamino)oxetane, 95%
Thermo Scientific Chemicals

3-(Methylamino)oxetane, 95%

CAS: 952182-03-5 | C4H9NO | 87.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57425.MD
also known as H57425-MD
Price (USD)
301.65
Special offer
Online exclusive
Ends: 30-Jun-2026
355.00
Save 53.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
301.65
Special offer
Online exclusive
Ends: 30-Jun-2026
355.00
Save 53.35 (15%)
Each
Chemical Identifiers
CAS952182-03-5
IUPAC NameN-methyloxetan-3-amine
Molecular FormulaC4H9NO
InChI KeyJUIXJPRSYHSLHK-UHFFFAOYSA-N
SMILESCNC1COC1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms
Appearance (Color)Colorless
Assay from Supplier's CofA≥94.0%
FormLiquid.
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Very Soluble (307 g/L) (25°C)

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Colin B. Reese.; Chamakura V. N. S. Varaprasad. Synthesis of some 2',3'-didehydro-2',3'-dideoxynucleosides derived from modified pyrimidine bases. J. Chem. Soc., Perkin Trans. 1994, 1 (2), 189-195.
  2. C. Richard Hall.; Thomas D. Inch. Methyl 4,6-anhydro-4-deoxy-2,3-di-O-methyl-4-methyl-amino-α-d-galactopyranoside and related compounds. Carbohydr. Res. 1977, 53 (2),254-258.