3-Bromooxetane, 95%
3-Bromooxetane, 95%
3-Bromooxetane, 95%
Thermo Scientific Chemicals

3-Bromooxetane, 95%

CAS: 39267-79-3 | C3H5BrO | 136.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57437.MD
also known as H57437-MD
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Specifications
CAS39267-79-3
Chemical Name or Material3-Bromooxetane
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.793
Flash Point48°C (118°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only