1-Oxa-6-azaspiro[3.4]octane oxalate, 96%
1-Oxa-6-azaspiro[3.4]octane oxalate, 96%
1-Oxa-6-azaspiro[3.4]octane oxalate, 96%
Thermo Scientific Chemicals

1-Oxa-6-azaspiro[3.4]octane oxalate, 96%

CAS: 1408074-51-0 | C8H13NO5 | 203.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57452.MD
also known as H57452-MD
Price (USD)
592.65
Online Exclusive
658.00
Save 65.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
592.65
Online Exclusive
658.00
Save 65.35 (10%)
Each
Specifications
CAS1408074-51-0
Chemical Name or Material1-Oxa-6-azaspiro[3.4]octane oxalate
Formula Weight203.19
Health Hazard 1H302+H312
Health Hazard 2GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
View more
1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only