Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57489.ME
also known as H57489-ME
500 mg
Catalog number H57489.ME
also known as H57489-ME
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Specifications
CAS100202-39-9
Chemical Name or MaterialMethyl azetidine-3-carboxylate hydrochloride
Flash Point69°C (156°F)
Formula Weight151.59
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only