1-Boc-3-methylazetidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-3-methylazetidine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57496.ME
also known as H57496-ME
500 mg
Catalog number H57496.ME
also known as H57496-ME
Price (USD)
297.65
Online Exclusive
331.00
Save 33.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
297.65
Online Exclusive
331.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS1314984-00-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown
Proton NMRConforms
Assay from Supplier's CofA≥94.0%
FormLiquid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only