2-Oxa-6-azaspiro[3.4]octane, 95%
2-Oxa-6-azaspiro[3.4]octane, 95%
2-Oxa-6-azaspiro[3.4]octane, 95%
2-Oxa-6-azaspiro[3.4]octane, 95%
Thermo Scientific Chemicals

2-Oxa-6-azaspiro[3.4]octane, 95%

CAS: 220290-68-6 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57579.ME
also known as H57579-ME
Price (USD)
469.65
Online Exclusive
522.00
Save 52.35 (10%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
469.65
Online Exclusive
522.00
Save 52.35 (10%)
Each
Add to cart
Specifications
CAS220290-68-6
Chemical Name or Material2-Oxa-6-azaspiro[3.4]octane
Density1.08
Flash Point63°C (145°F)
Formula Weight113.16
View more
2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents and bases.
RUO – Research Use Only
Zhao, F.; Lin, Z.; Wang, F.; Zhao, W.; Dong, X. Four-membered heterocycles-containing 4-anilino-quinazoline derivatives as epidermal growth factor receptor (EGFR) kinase inhibitors. Bioorg. Med. Chem. Lett. 2013, 23 (19), 5385-5388.