3-Amino-3-methyloxetane, 95%
3-Amino-3-methyloxetane, 95%
3-Amino-3-methyloxetane, 95%
Thermo Scientific Chemicals

3-Amino-3-methyloxetane, 95%

CAS: 874473-14-0 | C4H9NO | 87.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57588.MD
also known as H57588-MD
Price (USD)
172.65
Special offer
Online exclusive
Ends: 30-Jun-2026
203.00
Save 30.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
172.65
Special offer
Online exclusive
Ends: 30-Jun-2026
203.00
Save 30.35 (15%)
Each
Chemical Identifiers
CAS874473-14-0
IUPAC Name3-methyloxetan-3-amine
Molecular FormulaC4H9NO
InChI KeyNQVWMPOQWBDSAI-UHFFFAOYSA-N
SMILESCC1(N)COC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
CommentSolidifies below 0?C
Assay (GC)>94.0%
FormLiquid
It is used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only