1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals
1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals
1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57633.03
also known as H57633-03
1 g
Catalog number H57633.03
also known as H57633-03
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
120.00
Save 17.35 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
120.00
Save 17.35 (14%)
Each
Specifications
CAS34770-60-0
Chemical Name or Material1-Methyl-3-piperazinone
Formula Weight114.15
Health Hazard 1H302-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
View more
It is a useful piperazine and also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful piperazine and also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only