(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine, 95%
(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine, 95%
(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine, 95%
Thermo Scientific Chemicals

(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine, 95%

CAS: 22795-97-7 | C7H16N2 | 128.219 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H57638.03
also known as H57638-03
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
101.00
Save 15.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
101.00
Save 15.35 (15%)
Each
Specifications
CAS22795-97-7
Chemical Name or Material(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine
DOT InformationHazard Class: 8; Packaging Group: I
Density0.929
Flash Point57°C (135°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only