Chemical Name or Material(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine
DOT InformationHazard Class: 8; Packaging Group: I
Density0.929
Flash Point57°C (135°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.