1-Benzyl-5-bromoindole, 97%
1-Benzyl-5-bromoindole, 97%
1-Benzyl-5-bromoindole, 97%
1-Benzyl-5-bromoindole, 97%
Thermo Scientific Chemicals

1-Benzyl-5-bromoindole, 97%

CAS: 10075-51-1 | C15H12BrN | 286.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H57676.06
also known as H57676-06
Price (USD)
232.65
Special offer
Online exclusive
Ends: 30-Jun-2026
274.00
Save 41.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
232.65
Special offer
Online exclusive
Ends: 30-Jun-2026
274.00
Save 41.35 (15%)
Each
Add to cart
Specifications
CAS10075-51-1
Chemical Name or Material1-Benzyl-5-bromoindole
Formula Weight286.17
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only