3-Methoxyazetidine hydrochloride, 95%
3-Methoxyazetidine hydrochloride, 95%
3-Methoxyazetidine hydrochloride, 95%
3-Methoxyazetidine hydrochloride, 95%
Thermo Scientific Chemicals

3-Methoxyazetidine hydrochloride, 95%

CAS: 148644-09-1 | C4H10ClNO | 123.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57769.MD
also known as H57769-MD
Price (USD)
63.90
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
63.90
Each
Add to cart
Specifications
CAS148644-09-1
Chemical Name or Material3-Methoxyazetidine hydrochloride
Formula Weight123.58
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
RUO – Research Use Only

General References:

  1. Arthur G. Anderson.; Roger. Lok. Synthesis of azetidine-3-carboxylic acid.J. Org. Chem. 1972, 37 (24),3953-3955.
  2. Sonja Stanković.; Matthias D’hooghe.; Jo Dewulf.; Piet Bogaert.; Robrecht Jolie.; Norbert De Kimpe. Synthesis and reactivity of non-activated 2-(chloromethyl)aziridines.Tetrahedron Lett. 2011, 52 (35),4529-4532.