2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57776.ME
also known as H57776-ME
500 mg
Catalog number H57776.ME
also known as H57776-ME
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1158098-73-7
IUPAC Name2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
Molecular FormulaC7H13NO2S
InChI KeyVKUZMNXQGKBLHN-UHFFFAOYNA-N
SMILESCC(C)(C)S(=O)N=C1COC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow.
FormLow melting point solid.
Assay (HPLC)≥94.0%
Proton NMRConforms
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (1.00E+06 mg/L).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
RUO – Research Use Only