3-Azetidinemethanol hydrochloride, 95%
3-Azetidinemethanol hydrochloride, 95%
3-Azetidinemethanol hydrochloride, 95%
Thermo Scientific Chemicals

3-Azetidinemethanol hydrochloride, 95%

CAS: 928038-44-2 | C4H10ClNO | 123.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57801.MD
also known as H57801-MD
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.90
Save 14.25 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.90
Save 14.25 (15%)
Each
Chemical Identifiers
CAS928038-44-2
IUPAC Name(azetidin-3-yl)methanol hydrochloride
Molecular FormulaC4H10ClNO
InChI KeyAQUVQGSNKVDBBF-UHFFFAOYSA-N
SMILESCl.OCC1CNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
Proton NMRConforms to structure
3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.

Notes
Hygroscopic. Store at room temperature.
RUO – Research Use Only
Rama Subba Rao Vidadala.; Kayode K. Ojo.; Steven M. Johnson.; Zhongsheng Zhang.; Stephen E. Leonard.; Arinjay Mitra.; Ryan Choi.; Molly C. Reid.; Katelyn R. Keyloun.; Anna M.W. Fox.; Mark Kennedy.; Tiffany Silver-Brace.; Jen C.C. Hume.; Stefan Kappe.; Christophe L.M.J. Verlinde.; Erkang Fan.; Ethan A. Merritt.; Wesley C. Van Voorhis.; Dustin J. Maly. Development of potent and selective Plasmodium falciparum calcium-dependent protein kinase 4 (PfCDPK4) inhibitors that block the transmission of malaria to mosquitoes. Eur. J. Med. Chem. 2014, 74 562-573.