Thermo Scientific Chemicals

3-Oxetanone, 95%, Thermo Scientific Chemicals

Catalog number: H57870.03
1 g, Each
Thermo Scientific Chemicals

3-Oxetanone, 95%, Thermo Scientific Chemicals

Catalog number: H57870.03
1 g, Each
Quantity
Catalog number: H57870.03
also known as H57870-03
Price (USD)
Price: 151.00
Online price: 130.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6704-31-0
IUPAC Name
oxetan-3-one
Molecular Formula
C3H4O2
InChI Key
ROADCYAOHVSOLQ-UHFFFAOYSA-N
SMILES
O=C1COC1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
>94.0%

Description

3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents. Heat sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text