D-beta-Prolinol, 95%
D-beta-Prolinol, 95%
D-beta-Prolinol, 95%
Thermo Scientific Chemicals

D-beta-Prolinol, 95%

CAS: 110013-18-8 | C5H11NO | 101.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H57871.03
also known as H57871-03
Price (USD)
342.65
Online Exclusive
380.00
Save 37.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
342.65
Online Exclusive
380.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS110013-18-8
IUPAC Name(pyrrolidin-3-yl)methanol
Molecular FormulaC5H11NO
InChI KeyQOTUIIJRVXKSJU-UHFFFAOYNA-N
SMILESOCC1CCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow.
FormFused solid
Assay (GC)≥94.0%
Proton NMRConforms
Optical Rotation|b = +21? ?3? (C=0.8, CHCl3)
It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store at room temperature.
RUO – Research Use Only
David Alker.; Kevin J. Doyle.; Laurence M. Harwood.; Andrew McGregor. The direct synthesis of the cyclic sulphamidate of (S)-prolinol: SimultaneousN-protection and activation towards nucleophilic displacement of oxygen. Tetrahedron: Asymmetry. 1990, 1 (12),877-880.