(R)-(-)-2-Methylpyrrolidine, 95%
(R)-(-)-2-Methylpyrrolidine, 95%
(R)-(-)-2-Methylpyrrolidine, 95%
Thermo Scientific Chemicals

(R)-(-)-2-Methylpyrrolidine, 95%

CAS: 41720-98-3 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H57885.MD
also known as H57885-MD
Price (USD)
51.10
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
51.10
Each
Specifications
CAS41720-98-3
ChEBICHEBI:78858
Chemical Name or Material(R)-(-)-2-Methylpyrrolidine
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Density0.84
View more
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
RUO – Research Use Only

General References:

  1. Dalian Zhao.; Jeffrey T. Kuethe.; Michel Journet.; Zhihui Peng.; Guy R. Humphrey. Efficient and Practical Synthesis of (R)-2-Methylpyrrolidine.J. Org. Chem. 2006, 71 (11),4336-4338.
  2. Rumiko Sakurai.; Atsushi Yuzawa.; Kenichi Sakai.; Noriaki Hirayama. Dielectrically Controlled Enantiomeric Resolution (DCR) of (R)- and (S)-2-Methylpyrrolidine by (R,R)-Tartaric Acid.Dielectrically Controlled Enantiomeric Resolution (DCR) of (R)- and (S)-2-Methylpyrrolidine by (R,R)-Tartaric Acid. 2006, 6 (7),1606-1610.