Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals
Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals
Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57906.03
also known as H57906-03
1 g
Catalog number H57906.03
also known as H57906-03
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS36821-26-8
IUPAC Nameethyl 2-oxopyrrolidine-3-carboxylate
Molecular FormulaC7H11NO3
InChI KeyFHHMWJMUMOYGQW-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (GC)≥94.0%
Melting point69-75°C
Proton NMRConforms to structure
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Recommended storage temperature: 2 - 8°C Handle and store under inert gas.
RUO – Research Use Only
Toshiaki Suzuki.; Tomomi Goto.; Yoshitaka Hamashima.; Mikiko Sodeoka. Enantioselective Fluorination of tert-Butoxycarbonyl Lactones and Lactams Catalyzed by Chiral Pd(II)-Bisphosphine Complexes. J. Org. Chem. 2007, 72 (1),246-250.