Benzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Benzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Benzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Benzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 62673-31-8 | C7H7BrZn | 236.417 g/mol
Have Questions?
Catalog NumberQuantity
H58014.AD
also known as H58014-AD
50 mL
Catalog number H58014.AD
also known as H58014-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS62673-31-8
IUPAC Namebromozincylium; phenylmethanide
Molecular FormulaC7H7BrZn
InChI KeyWRSWIWOVJBYZAW-UHFFFAOYSA-M
SMILES[Zn+]Br.[CH2-]C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)0.4-0.6M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in cross coupling reactions.

Solubility
Reacts with water.

Notes
Air sensitive. Recommended storage temperature: 2 - 8°C.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Ming Li.; Xiao-Xia Yan.; Wei Hong.; Xia-Zhen Zhu.; Bo-Xun Cao.; Jie Sun.; Xue-Long Hou. Palladium-Catalyzed Enantioselective Ring Opening of Oxabicyclic Alkenes with Organozinc Halides. Org. Lett. 2004, 6 (16),2833-2835.