4-(Maleimidomethyl)benzonitrile, 97%, Thermo Scientific Chemicals
4-(Maleimidomethyl)benzonitrile, 97%, Thermo Scientific Chemicals
4-(Maleimidomethyl)benzonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Maleimidomethyl)benzonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H58015.03
also known as H58015-03
Price (USD)
164.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
164.00
Each
Chemical Identifiers
CAS1249094-81-2
IUPAC Name4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]benzonitrile
Molecular FormulaC12H8N2O2
InChI KeyDFJVGKSXXPQRLG-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)N1CC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
Assay (GC)>96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only