Isoxazole-3-carboxylic acid, 97%
Isoxazole-3-carboxylic acid, 97%
Isoxazole-3-carboxylic acid, 97%
Thermo Scientific Chemicals

Isoxazole-3-carboxylic acid, 97%

CAS: 3209-71-0 | C4H3NO3 | 113.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H58438.03
also known as H58438-03
Price (USD)
148.65
Online exclusive
165.00
Save 16.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
148.65
Online exclusive
165.00
Save 16.35
Each
Specifications
Beilstein110919
CAS3209-71-0
Chemical Name or MaterialIsoxazole-3-carboxylic acid
Formula Weight113.07
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only