4-Chlorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-Chlorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-Chlorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

4-Chlorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 149923-10-4 | C7H6Cl2Zn | 226.41 g/mol
Have Questions?
Catalog NumberQuantity
H58532.AD
also known as H58532-AD
50 mL
Catalog number H58532.AD
also known as H58532-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS149923-10-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Concentration0.40-0.60M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorobenzylzinc chloride used as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Y. Fu; M. Yuan; X. Hu; Y. Yang; and H. Hou. 5-{2-(4-Chloro­phen­yl)-1-[2-(4-chloro­phen­yl)-1-(3,4,5-trimeth­oxy­phen­yl)eth­oxy]eth­yl}-1,2,3-trimeth­oxy­benzene. Acta Cryst.,2012, E68, o1007.
  2. Zhi-Yong Peng; Fang-Fang Ma; Lv-Feng Zhu; Xiao-Min Xie; and Zhaoguo Zhang. Lewis Acid Promoted Carbon-Carbon Double-Bond Formation via Organozinc Reagents and Carbonyl Compounds. J. Org. Chem. 2009, 74 (17), 6855-6858.