Isopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Isopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Isopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Isopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 77047-87-1 | C3H7BrZn | 188.37 g/mol
Have Questions?
Catalog NumberQuantity
H58536.AD
also known as H58536-AD
50 mL
Catalog number H58536.AD
also known as H58536-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS77047-87-1
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.4-0.6M in THF
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropylzinc bromide is employed in croos-coupling reactions.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents,
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Chong Han.; Stephen L. Buchwald. Negishi Coupling of Secondary Alkylzinc Halides with Aryl Bromides and Chlorides. J. Am. Chem. Soc. 2009, 131 (22),7532-7533.