2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals
2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals
2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H58554.04
also known as H58554-04
2 g
Catalog number H58554.04
also known as H58554-04
Price (USD)
259.65
Special offer
Online exclusive
Ends: 30-Jun-2026
305.00
Save 45.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
259.65
Special offer
Online exclusive
Ends: 30-Jun-2026
305.00
Save 45.35 (15%)
Each
Specifications
CAS30235-26-8
Chemical Name or Material2-Amino-4-(2-pyridyl)thiazole
ColorBrown
Formula Weight177.22
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only