(R)-(+)-1,2-Epoxybutane, 98%
(R)-(+)-1,2-Epoxybutane, 98%
(R)-(+)-1,2-Epoxybutane, 98%
(R)-(+)-1,2-Epoxybutane, 98%
Thermo Scientific Chemicals

(R)-(+)-1,2-Epoxybutane, 98%

CAS: 3760-95-0 | C4H8O | 72.107 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H58579.03
also known as H58579-03
Price (USD)
57.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.00
Each
Add to cart
Specifications
CAS3760-95-0
Chemical Name or Material(R)-(+)-1,2-Epoxybutane
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-BUTYLENE OXIDE, STABILIZED
Density0.83
Flash Point−12°C (10°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only