4-Fluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-Fluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-Fluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

4-Fluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 181705-93-1 | C6H4BrFZn | 240.38 g/mol
Have Questions?
Catalog NumberQuantity
H58780.AD
also known as H58780-AD
50 mL
Catalog number H58780.AD
also known as H58780-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
CAS181705-93-1
Chemical Name or Material4-Fluorophenylzinc bromide
DOT InformationTransport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Density0.992
Formula Weight240.38
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only