2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester, 96%
2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester, 96%
2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester, 96%
Thermo Scientific Chemicals

2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester, 96%

CAS: 303006-90-8 | C18H26B2F2O4 | 366.019 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59025.03
also known as H59025-03
Price (USD)
323.65
Online Exclusive
359.00
Save 35.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
323.65
Online Exclusive
359.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS303006-90-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
Assay from Supplier's CofA≥95.0%
FormPowder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only