2-Amino-4-chloropyridine, 97%
2-Amino-4-chloropyridine, 97%
2-Amino-4-chloropyridine, 97%
Thermo Scientific Chemicals

2-Amino-4-chloropyridine, 97%

CAS: 19798-80-2 | C5H6ClN2 | 129.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H59130.06
also known as H59130-06
Price (USD)
56.65
Online Exclusive
63.00
Save 6.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.00
Save 6.35 (10%)
Each
Chemical Identifiers
CAS19798-80-2
IUPAC Name2-amino-4-chloropyridin-1-ium
Molecular FormulaC5H6ClN2
InChI KeyRQMWVVBHJMUJNZ-UHFFFAOYSA-O
SMILESNC1=CC(Cl)=CC=[NH+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Assay (HPLC)>96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only