Specifications
CAS775351-40-1
Chemical Name or Material4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester
Formula Weight272.11
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more