Specifications
CAS261732-38-1
Chemical Name or Material1-Boc-5-bromoindoline
Formula Weight298.18
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more