4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals
4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals
4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H59397.03
also known as H59397-03
1 g
Catalog number H59397.03
also known as H59397-03
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
Specifications
CAS330794-03-1
Chemical Name or Material4-Bromo-5-fluoro-2-methoxyaniline
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight220.04
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.

Notes
Store in a cool place. Keep the container tightly closed. Incompatible with strong-oxidizing agents, strong acids and strong bases.
RUO – Research Use Only