1-Boc-indoline-5-boronic acid pinacol ester, 97%
1-Boc-indoline-5-boronic acid pinacol ester, 97%
1-Boc-indoline-5-boronic acid pinacol ester, 97%
1-Boc-indoline-5-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

1-Boc-indoline-5-boronic acid pinacol ester, 97%

CAS: 837392-67-3 | C19H28BNO4 | 345.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59436.03
also known as H59436-03
Price (USD)
260.65
Online Exclusive
289.00
Save 28.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
260.65
Online Exclusive
289.00
Save 28.35 (10%)
Each
Add to cart
Specifications
CAS837392-67-3
Chemical Name or Material1-Boc-indoline-5-boronic acid pinacol ester
Formula Weight345.25
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only