3-Fluoroazetidine hydrochloride, 95%
3-Fluoroazetidine hydrochloride, 95%
3-Fluoroazetidine hydrochloride, 95%
Thermo Scientific Chemicals

3-Fluoroazetidine hydrochloride, 95%

CAS: 617718-46-4 | C3H7ClFN | 111.544 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H59454.ME
also known as H59454-ME
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Specifications
CAS617718-46-4
Chemical Name or Material3-Fluoroazetidine hydrochloride
Formula Weight111.55
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Eva Van Hende.; Guido Verniest.; Frederik Deroose.; Jan-Willem Thuring.; Gregor Macdonald.; Norbert De Kimpe. Synthesis of 1-Boc-3-fluoroazetidine-3-carboxylic Acid. J. Org. Chem. 2009, 74 (5),2250-2253.
  2. Willem Van Brabandt.; Guido Verniest.; Dirk De Smaele.; Guillaume Duvey.; Norbert De Kimpe. Synthesis of 3-Fluoroazetidines. J. Org. Chem. 2006, 71 (18),7100-7102.