1-Ethyl-3-methylimidazolium methyl sulfate, 98%
1-Ethyl-3-methylimidazolium methyl sulfate, 98%
1-Ethyl-3-methylimidazolium methyl sulfate, 98%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium methyl sulfate, 98%

CAS: 516474-01-4 | C7H14N2O4S | 222.259 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H59480.18
also known as H59480-18
Price (USD)
186.65
Special offer
Online exclusive
Ends: 30-Jun-2026
219.00
Save 32.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
186.65
Special offer
Online exclusive
Ends: 30-Jun-2026
219.00
Save 32.35 (15%)
Each
Chemical Identifiers
CAS516474-01-4
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium methyl sulfate
Molecular FormulaC7H14N2O4S
InChI KeyBXSDLSWVIAITRQ-UHFFFAOYSA-M
SMILESCOS([O-])(=O)=O.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only