1-n-Hexyl-3-methylimidazolium iodide, 98%
1-n-Hexyl-3-methylimidazolium iodide, 98%
1-n-Hexyl-3-methylimidazolium iodide, 98%
Thermo Scientific Chemicals

1-n-Hexyl-3-methylimidazolium iodide, 98%

CAS: 178631-05-5 | C10H19IN2 | 294.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H59487.06
also known as H59487-06
Price (USD)
41.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.40
Each
Chemical Identifiers
CAS178631-05-5
IUPAC Name1-hexyl-3-methyl-1H-imidazol-3-ium iodide
Molecular FormulaC10H19IN2
InChI KeyCZIUVCSYOGFUPH-UHFFFAOYSA-M
SMILES[I-].CCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only