3,4-Dichloro-6-fluoroaniline, 96%
3,4-Dichloro-6-fluoroaniline, 96%
3,4-Dichloro-6-fluoroaniline, 96%
3,4-Dichloro-6-fluoroaniline, 96%
Thermo Scientific Chemicals

3,4-Dichloro-6-fluoroaniline, 96%

CAS: 2729-36-4 | C6H4Cl2FN | 180.003 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59506.03
also known as H59506-03
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Add to cart
Specifications
CAS2729-36-4
Chemical Name or Material3,4-Dichloro-6-fluoroaniline
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight180.01
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
3,4-Dichloro-6-fluoroaniline is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichloro-6-fluoroaniline is used as pharmaceutical intermediates

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Iris Ben-David.; Yulia Rozen.; Giuseppina Ortu, Eyal Mishani. Radiosynthesis of ML03, a novel positron emission tomography biomarker for targeting epidermal growth factor receptor via the labeling synthon: [11C]acryloyl chloride. Applied Radiation and Isotopes. 2003, 58, (2), 209-217.