Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%
Thermo Scientific Chemicals

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%

CAS: 2389-45-9 | C19H28N2O6 | 380.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number H59537.04
also known as H59537-04
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS2389-45-9
IUPAC Name(2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Molecular FormulaC19H28N2O6
InChI KeyBDHUTRNYBGWPBL-GGYSOQFKNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only