Specifications
CAS1337596-50-5
Chemical Name or Material6-Amino-7-(5-indolinyl)-9-methyl-7-deazapurine
Formula Weight265.31
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more