2-Bromo-2',4'-dichloroacetophenone, 95%
2-Bromo-2',4'-dichloroacetophenone, 95%
2-Bromo-2',4'-dichloroacetophenone, 95%
Thermo Scientific Chemicals

2-Bromo-2',4'-dichloroacetophenone, 95%

CAS: 2631-72-3 | C8H5BrCl2O | 267.931 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H59615.03
also known as H59615-03
Price (USD)
69.65
Online Exclusive
77.70
Save 8.05 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.70
Save 8.05 (10%)
Each
Chemical Identifiers
CAS2631-72-3
IUPAC Name2-bromo-1-(2,4-dichlorophenyl)ethan-1-one
Molecular FormulaC8H5BrCl2O
InChI KeyDASJDMQCPIDJIF-UHFFFAOYSA-N
SMILESClC1=CC=C(C(=O)CBr)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only